Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EHF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206615.1 | NP_001193544.1 | 3 | 455 | ||
NM_001206616.1 | NP_001193545.1 | 3 | 456 | |||
NM_012153.5 | NP_036285.2 | 3 | 455 | |||
XM_005252857.1 | XP_005252914.1 | 3 | 456 | |||
XM_005252860.1 | XP_005252917.1 | 3 | 309 | |||
XM_005252861.1 | XP_005252918.1 | 3 | 456 | |||
XM_005252862.1 | XP_005252919.1 | 3 | 325 | |||
XM_011519983.1 | XP_011518285.1 | 3 | 481 | |||
XM_011519984.1 | XP_011518286.1 | 4 | 561 | |||
XM_011519985.1 | XP_011518287.1 | 3 | 346 | |||
XM_017017534.1 | XP_016873023.1 | 3 | 456 | |||
GenBank mRNA | AF203977.1 | - | 3 | 376 | ||
AF212848.1 | - | 3 | 395 | |||
AK310867.1 | - | 3 | 456 | |||
BC038995.2 | - | 3 | 415 | 6055934 | ||
DA460976.1 | - | 3 | 454 | |||
DB214606.1 | - | 4 | 346 |