Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EHF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206616.1 | NP_001193545.1 | 5, 6 | 657 | ||
NM_012153.5 | NP_036285.2 | 5, 6 | 656 | |||
XM_005252857.1 | XP_005252914.1 | 5, 6 | 657 | |||
XM_005252860.1 | XP_005252917.1 | 5, 6 | 510 | |||
XM_011519983.1 | XP_011518285.1 | 5, 6 | 682 | |||
XM_011519984.1 | XP_011518286.1 | 6, 7 | 762 | |||
XM_011519985.1 | XP_011518287.1 | 5, 6 | 547 | |||
GenBank mRNA | AF124439.1 | - | 5, 6 | 562 | ||
AF170583.1 | - | 5, 6 | 566 | |||
AF203977.1 | - | 5, 6 | 577 | |||
AF212848.1 | - | 5, 6 | 596 | |||
AK309318.1 | - | 5, 6 | 682 | |||
AK310867.1 | - | 5, 6 | 657 | |||
AY882601.1 | - | 4, 5 | 465 | |||
BC038995.2 | - | 5, 6 | 616 | 6055934 |