Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COPG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290033.1 | NP_001276962.1 | 11, 12 | 1026 | ||
NM_012133.5 | NP_036265.3 | 11, 12 | 1026 | |||
GenBank mRNA | AB047847.1 | - | 11, 12 | 936 | ||
AF157833.1 | - | 11, 12 | 996 | |||
AF207598.1 | - | 11, 12 | 992 | |||
AK002086.1 | - | 3, 4 | 328 | |||
AK022934.1 | - | 11, 12 | 923 | |||
AK024414.1 | - | 4, 5 | 404 | |||
AK291409.1 | - | 11, 12 | 1016 | |||
BC017443.1 | - | 11, 12 | 919 | |||
BC110796.1 | - | 11, 12 | 973 | 5197024 |