Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKAP8L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014371.3 | NP_055186.3 | 4 | 486 | ||
XM_011527916.2 | XP_011526218.1 | 4 | 393 | |||
GenBank mRNA | AB015332.1 | - | 4 | 333 | ||
AB025905.1 | - | 4 | 333 | |||
AB451326.1 | - | 3 | 288 | |||
AB451469.1 | - | 3 | 288 | |||
AJ243467.1 | - | 4 | 334 | |||
AK001594.1 | - | 4 | 353 | |||
AK225046.1 | - | 4 | 353 | |||
AK225291.1 | - | 4 | 353 | |||
BC000713.2 | - | 4 | 338 | 3504388 | ||
BI458860.1 | - | 4 | 391 |