Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRUB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015679.1 | NP_056494.1 | 6, 7 | 550 | ||
GenBank mRNA | AF131848.1 | - | 6, 7 | 562 | ||
AJ420462.1 | - | 6, 7 | 554 | |||
AK001956.1 | - | 6, 7 | 550 | |||
AK301986.1 | - | 6, 7 | 795 | |||
BC001457.2 | - | 6, 7 | 569 | 3138788 |