Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYFIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037333.2 | NP_001032410.1 | 12 | 1335 | ||
NM_001291721.1 | NP_001278650.1 | 11 | 1257 | |||
NM_001291722.1 | NP_001278651.1 | 12 | 1335 | |||
NM_014376.3 | NP_055191.2 | 12 | 1461 | |||
XM_011534516.2 | XP_011532818.1 | 12 | 1601 | |||
XM_017009341.1 | XP_016864830.1 | 12 | 1450 | |||
XM_017009342.1 | XP_016864831.1 | 13 | 1372 | |||
GenBank mRNA | AB032994.2 | - | 12 | 1275 | ||
AF160973.1 | - | 12 | 1244 | |||
AK223604.1 | - | 11 | 1213 | |||
AK294220.1 | - | 7 | 704 | |||
AK295865.1 | - | 9 | 884 | |||
AK297401.1 | - | 10 | 1067 | |||
AK303702.1 | - | 11 | 1210 | |||
AL136549.1 | - | 12 | 1334 | |||
AL161999.1 | - | 3 | 220 | |||
BC011762.1 | - | 12 | 1461 | 3619680 | ||
BC021008.1 | - | 12 | 1461 | |||
L47738.1 | - | 1 | 54 |