Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014443.2 | NP_001014443.1 | 13 | 1962 | ||
NM_001319848.1 | NP_001306777.1 | 12 | 1779 | |||
NM_012475.4 | NP_036607.3 | 12 | 1821 | |||
XM_017001036.1 | XP_016856525.1 | 11 | 2106 | |||
XM_017001037.1 | XP_016856526.1 | 12 | 2160 | |||
XM_017001038.1 | XP_016856527.1 | 12 | 2196 | |||
XM_017001039.1 | XP_016856528.1 | 11 | 2049 | |||
XM_017001040.1 | XP_016856529.1 | 12 | 1884 | |||
XM_017001041.1 | XP_016856530.1 | 12 | 1845 | |||
XM_017001042.1 | XP_016856531.1 | 7 | 1004 | |||
GenBank mRNA | AB208899.1 | - | 8 | 2738 | ||
AF177758.1 | - | 12 | 1672 | |||
AF217979.1 | - | 12 | 1703 | |||
AF233442.1 | - | 12 | 1759 | |||
AI038040.1 | - | 2 | 393 | |||
AK292319.1 | - | 13 | 1907 | |||
AK310326.1 | - | 12 | 1854 | |||
AL157417.1 | - | 8 | 2137 | |||
BC003130.1 | - | 11 | 1321 | |||
BC090946.1 | - | 13 | 1910 | 3877298 | ||
BC136291.1 | - | 11 | 1609 | 9020278 |