Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SND1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014390.2 | NP_055205.2 | 9, 10 | 1254 | ||
XM_017011987.1 | XP_016867476.1 | 9, 10 | 1283 | |||
GenBank mRNA | AB209510.1 | - | 9, 10 | 1192 | ||
AK096583.1 | - | 10, 11 | 1179 | |||
AK304181.1 | - | 9, 10 | 1146 | |||
AK312313.1 | - | 9, 10 | 1208 | |||
BC017180.2 | - | 9, 10 | 1210 | 3345037 | ||
BT009785.1 | - | 9, 10 | 1028 | |||
U22055.1 | - | 9, 10 | 1220 |