Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TUBGCP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286414.2 | NP_001273343.1 | 14 | 1850 | ||
NM_014444.4 | NP_055259.2 | 14 | 1847 | |||
XM_011521454.2 | XP_011519756.1 | 14 | 1850 | |||
XM_011521455.2 | XP_011519757.1 | 14 | 1748 | |||
XM_017022078.1 | XP_016877567.1 | 14 | 1847 | |||
GenBank mRNA | AJ249677.1 | - | 14 | 1556 | ||
AK001639.1 | - | 10 | 1161 | |||
AK026551.1 | - | 14 | 1837 | |||
AK027703.1 | - | 14 | 1808 | |||
AK127306.1 | - | 4 | 1991 | |||
BC000966.2 | - | 13 | 1373 | 3446803 | ||
BC009870.2 | - | 14 | 1772 | 3940649 | ||
BC012801.2 | - | 14 | 1791 | 2821891 |