Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SULT1B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014465.3 | NP_055280.2 | 4, 5 | 679 | ||
XM_005265676.3 | XP_005265733.1 | 5, 6 | 594 | |||
XM_005265677.3 | XP_005265734.1 | 5, 6 | 794 | |||
XM_005265678.3 | XP_005265735.1 | 4, 5 | 547 | |||
XM_011531873.2 | XP_011530175.1 | 3, 4 | 418 | |||
GenBank mRNA | BC010895.1 | - | 4, 5 | 681 | 4105139 | |
D89479.1 | - | 3, 4 | 405 | |||
U95726.1 | - | 3, 4 | 396 |