Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF330 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014487.5 | NP_055302.1 | 3, 4 | 376 | ||
XM_017008033.1 | XP_016863522.1 | 3, 4 | 520 | |||
GenBank mRNA | AJ006591.2 | - | 3, 4 | 321 | ||
AK002106.1 | - | 3, 4 | 326 | |||
AK223609.1 | - | 3, 4 | 327 | |||
AK315463.1 | - | 3, 4 | 362 | |||
BC004421.1 | - | 3, 4 | 352 | 3352916 | ||
DA089047.1 | - | 3, 4 | 368 |