Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDH19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021153.3 | NP_066976.1 | 11 | 1937 | ||
NR_073130.1 | 11 | 1909 | ||||
XM_011525931.2 | XP_011524233.1 | 11 | 1904 | |||
XM_017025711.1 | XP_016881200.1 | 11 | 1826 | |||
GenBank mRNA | AF047826.1 | - | 3 | 275 | ||
AJ007607.1 | - | 11 | 1778 | |||
AK315257.1 | - | 11 | 1911 | |||
AY358654.1 | - | 10 | 1676 |