Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDH19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271028.1 | NP_001257957.1 | 4, 5 | 908 | ||
NM_021153.3 | NP_066976.1 | 4, 5 | 908 | |||
NR_073130.1 | 4, 5 | 908 | ||||
XM_011525931.2 | XP_011524233.1 | 4, 5 | 875 | |||
XM_011525932.1 | XP_011524234.1 | 3, 4 | 686 | |||
XM_017025711.1 | XP_016881200.1 | 4, 5 | 797 | |||
GenBank mRNA | AJ007607.1 | - | 4, 5 | 749 | ||
AK304000.1 | - | 4, 5 | 858 | |||
AK315257.1 | - | 4, 5 | 910 | |||
AY358654.1 | - | 3, 4 | 647 | |||
BC015877.1 | - | 4, 5 | 800 | 4716394 |