Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C11orf54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286067.1 | NP_001272996.1 | 7 | 710 | ||
NM_001286068.1 | NP_001272997.1 | 6 | 573 | |||
NM_001286069.1 | NP_001272998.1 | 6 | 578 | |||
NM_001286070.1 | NP_001272999.1 | 5 | 426 | |||
NM_014039.3 | NP_054758.2 | 6 | 578 | |||
XM_006718824.1 | XP_006718887.1 | 7 | 619 | |||
XM_011542781.2 | XP_011541083.1 | 6 | 892 | |||
XM_011542782.2 | XP_011541084.1 | 6 | 983 | |||
XM_017017613.1 | XP_016873102.1 | 7 | 4034 | |||
XM_017017614.1 | XP_016873103.1 | 7 | 861 | |||
XM_017017615.1 | XP_016873104.1 | 7 | 1314 | |||
XM_017017616.1 | XP_016873105.1 | 6 | 570 | |||
XM_017017619.1 | XP_016873108.1 | 5 | 424 | |||
GenBank mRNA | AF092133.1 | - | 6 | 486 | ||
AK225977.1 | - | 6 | 508 | |||
AK291833.1 | - | 6 | 484 | |||
AK292215.1 | - | 7 | 629 | |||
AL136605.1 | - | 6 | 504 | |||
AY203960.1 | - | 5 | 453 | |||
BC007110.1 | - | 5 | 333 | |||
BC012298.1 | - | 6 | 545 | 3852558 | ||
BI597310.1 | - | 5 | 348 | |||
CR533538.1 | - | 5 | 346 |