Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM14A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014051.3 | NP_054770.1 | 2, 3 | 251 | ||
GenBank mRNA | AF078864.1 | - | 2, 3 | 295 | ||
AF239771.1 | - | 2, 3 | 230 | |||
AK312063.1 | - | 2, 3 | 224 | |||
BC015097.1 | - | 2, 3 | 210 | 3686196 | ||
BC019328.1 | - | 2, 3 | 206 | 3534851 | ||
CB117714.1 | - | 2, 3 | 251 |