Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014990.2 | NP_001014990.1 | 6 | 400 | ||
NM_014180.3 | NP_054899.2 | 7 | 518 | |||
GenBank mRNA | AF161507.1 | - | 7 | 486 | ||
AK000601.1 | - | 7 | 505 | |||
AK097880.1 | - | 3 | 2155 | |||
AK302784.1 | - | 6 | 380 | |||
BC012565.1 | - | 7 | 488 | 4333117 | ||
BM662504.1 | - | 1 | 226 | |||
BP372420.1 | - | 7 | 518 | |||
BU943770.1 | - | 7 | 511 | |||
CR933679.1 | - | 6 | 379 |