Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYLIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013262.3 | NP_037394.2 | 6 | 1165 | ||
XM_005249032.2 | XP_005249089.1 | 5 | 1000 | |||
XM_005249033.2 | XP_005249090.1 | 5 | 889 | |||
XM_017010789.1 | XP_016866278.1 | 6 | 1165 | |||
GenBank mRNA | AF006003.1 | - | 6 | 1078 | ||
AF187016.2 | - | 6 | 1102 | |||
AF258586.1 | - | 6 | 1142 | |||
AK026739.1 | - | 3 | 383 | |||
AK074391.1 | - | 6 | 1165 | |||
AK314648.1 | - | 6 | 1165 | |||
BC002860.2 | - | 6 | 1146 | 3638617 | ||
BT007055.1 | - | 6 | 928 |