Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323623.1 | NP_001310552.1 | 14 | 2018 | ||
NM_001323624.1 | NP_001310553.1 | 12 | 1629 | |||
NM_013285.2 | NP_037417.1 | 13 | 1814 | |||
GenBank mRNA | AK130697.1 | - | 1 | 1827 | ||
AK298358.1 | - | 13 | 1633 | |||
AK312915.1 | - | 13 | 1768 | |||
BC000107.2 | - | 13 | 1750 | 3506131 | ||
BC009250.2 | - | 13 | 1750 | 2960125 | ||
L05425.1 | - | 13 | 1730 |