Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRA2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282757.1 | NP_001269686.1 | 3, 4 | 698 | ||
NM_001282758.1 | NP_001269687.1 | 3, 4 | 487 | |||
NM_001282759.1 | NP_001269688.1 | 4, 5 | 788 | |||
NM_013293.4 | NP_037425.1 | 2, 3 | 397 | |||
XM_005249725.1 | XP_005249782.1 | 3, 4 | 684 | |||
XM_011515331.1 | XP_011513633.1 | 2, 3 | 388 | |||
XM_017012089.1 | XP_016867578.1 | 2, 3 | 929 | |||
XM_017012090.1 | XP_016867579.1 | 4, 5 | 739 | |||
GenBank mRNA | AB097048.1 | - | 2, 3 | 338 | ||
AK298815.1 | - | 4, 5 | 749 | |||
AK300565.1 | - | 3, 4 | 640 | |||
AK300741.1 | - | 3, 4 | 427 | |||
AK310719.1 | - | 2, 3 | 929 | |||
AK311687.1 | - | 3, 4 | 649 | |||
AK314801.1 | - | 2, 3 | 338 | |||
BC017094.2 | - | 2, 3 | 308 | 3907327 | ||
BG722500.1 | - | 2, 3 | 401 | |||
BP376263.1 | - | 2, 3 | 388 | |||
DB445598.1 | - | 2, 3 | 395 | |||
U53209.1 | - | 2, 3 | 337 |