Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C18orf8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318707.1 | NP_001305636.1 | 16 | 1620 | ||
NM_001318709.1 | NP_001305638.1 | 17 | 1705 | |||
NM_013326.4 | NP_037458.3 | 19 | 1849 | |||
XM_011525939.1 | XP_011524241.1 | 18 | 1721 | |||
XM_017025717.1 | XP_016881206.1 | 17 | 1658 | |||
GenBank mRNA | AF143536.1 | - | 18 | 1632 | ||
AK027106.1 | - | 11 | 925 | |||
AK057192.1 | - | 19 | 1810 | |||
AK294169.1 | - | 16 | 1567 | |||
AK296230.1 | - | 17 | 1656 | |||
BC002950.1 | - | 19 | 1774 | 3538017 | ||
BC008305.1 | - | 19 | 1781 | 3537084 |