Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NME7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013330.4 | NP_037462.1 | 8 | 1025 | ||
NM_197972.2 | NP_932076.1 | 8 | 994 | |||
NR_104229.1 | 8 | 1025 | ||||
XM_005245106.4 | XP_005245163.1 | 6 | 603 | |||
XM_017001099.1 | XP_016856588.1 | 8 | 1011 | |||
XM_017001100.1 | XP_016856589.1 | 6 | 591 | |||
GenBank mRNA | AF153191.2 | - | 8 | 864 | ||
AK094513.1 | - | 8 | 977 | |||
AK290701.1 | - | 8 | 859 | |||
AK301917.1 | - | 8 | 1008 | |||
BC006983.1 | - | 8 | 856 | 3997152 |