Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DSE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080976.2 | NP_001074445.1 | 3 | 736 | ||
NM_001322937.1 | NP_001309866.1 | 4 | 1292 | |||
NM_001322938.1 | NP_001309867.1 | 4 | 1204 | |||
NM_001322939.1 | NP_001309868.1 | 3 | 619 | |||
NM_001322940.1 | NP_001309869.1 | 4 | 1288 | |||
NM_001322941.1 | NP_001309870.1 | 3 | 786 | |||
NM_001322943.1 | NP_001309872.1 | 3 | 790 | |||
NM_001322944.1 | NP_001309873.1 | 3 | 790 | |||
NM_013352.3 | NP_037484.1 | 3 | 790 | |||
NR_136524.1 | 3 | 732 | ||||
XM_017010795.1 | XP_016866284.1 | 5 | 925 | |||
XM_017010796.1 | XP_016866285.1 | 2 | 546 | |||
XM_017010797.1 | XP_016866286.1 | 3 | 1470 | |||
GenBank mRNA | AF098066.1 | - | 3 | 634 | ||
AK301513.1 | - | 3 | 619 | |||
BC039245.1 | - | 3 | 681 | 5272885 |