Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199054.1 | NP_001185983.1 | 5 | 634 | ||
NM_001199055.1 | NP_001185984.1 | 5 | 485 | |||
NM_001199056.1 | NP_001185985.1 | 5 | 365 | |||
NM_013399.2 | NP_037531.2 | 5 | 602 | |||
GenBank mRNA | AF131218.1 | - | 4 | 681 | ||
AF195661.1 | - | 5 | 597 | |||
AK292036.1 | - | 5 | 560 | |||
AK294257.1 | - | 5 | 455 | |||
AK302297.1 | - | 5 | 323 | |||
AK307067.1 | - | 6 | 563 | |||
AK311295.1 | - | 1 | 58 | |||
AL136698.1 | - | 5 | 560 | |||
AL833853.1 | - | 5 | 634 | |||
BC002882.2 | - | 5 | 548 | 3941342 | ||
BC007604.1 | - | 1 | 136 | |||
CR533446.1 | - | 3 | 384 | |||
DQ167023.1 | - | 5 | 571 |