Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RHOD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300886.1 | NP_001287815.1 | 2 | 296 | ||
NM_014578.3 | NP_055393.1 | 4 | 494 | |||
GenBank mRNA | AF498973.1 | - | 4 | 409 | ||
BC001338.1 | - | 4 | 462 | 3461774 | ||
BM761362.1 | - | 4 | 494 | |||
CB853597.1 | - | 2 | 653 | |||
CN305061.1 | - | 2 | 225 |