Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LMCD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278234.1 | NP_001265163.1 | 2 | 209 | ||
NM_001278235.1 | NP_001265164.1 | 2 | 209 | |||
NM_014583.3 | NP_055398.1 | 2 | 209 | |||
XM_011533649.1 | XP_011531951.1 | 3 | 264 | |||
GenBank mRNA | AF169284.1 | - | 2 | 180 | ||
AF216709.1 | - | 2 | 98 | |||
AK022176.1 | - | 2 | 191 | |||
AK293849.1 | - | 2 | 130 | |||
AK300803.1 | - | 2 | 193 | |||
BC000646.2 | - | 2 | 179 | 3343074 | ||
CN388104.1 | - | 2 | 209 |