Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SOX8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014587.4 | NP_055402.2 | 1 | 358 | ||
GenBank mRNA | AF226675.1 | - | 1 | 243 | ||
AK024491.1 | - | 1 | 80 | |||
BC031797.1 | - | 1 | 358 | 4937883 |