Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNIP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034914.1 | NP_001030086.1 | 3 | 253 | ||
NM_013434.4 | NP_038462.1 | 4 | 446 | |||
GenBank mRNA | AF120102.1 | - | 4 | 341 | ||
AF199599.1 | - | 3 | 331 | |||
AJ131730.2 | - | 4 | 433 | |||
AK315437.1 | - | 4 | 392 | |||
BC012850.1 | - | 4 | 365 | 4179275 | ||
BI755669.1 | - | 4 | 490 | |||
BT020075.1 | - | 3 | 331 | |||
DQ148485.1 | - | 3 | 331 | |||
DQ148486.1 | - | 3 | 253 |