Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DERL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304777.1 | NP_001291706.1 | 5 | 492 | ||
NM_016041.4 | NP_057125.2 | 5 | 495 | |||
NR_130905.1 | 4 | 607 | ||||
NR_130906.1 | 4 | 401 | ||||
GenBank mRNA | AF132289.1 | - | 5 | 443 | ||
AF151859.1 | - | 5 | 433 | |||
AF208065.1 | - | 5 | 431 | |||
AF242523.1 | - | 5 | 431 | |||
AK024574.1 | - | 1 | 113 | |||
BC010890.1 | - | 5 | 443 | 4280273 | ||
BG110482.1 | - | 4 | 556 | |||
CN362881.1 | - | 5 | 413 | |||
CN362882.1 | - | 4 | 319 | |||
CR457202.1 | - | 5 | 426 | |||
HY051246.1 | - | 4 | 361 |