Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TXNDC11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303447.1 | NP_001290376.1 | 7, 8 | 1126 | ||
NM_001324022.1 | NP_001310951.1 | 5, 6 | 915 | |||
NM_015914.6 | NP_056998.4 | 6, 7 | 1045 | |||
NR_136671.1 | 6, 7 | 1045 | ||||
NR_136673.1 | 5, 6 | 915 | ||||
XM_011522515.2 | XP_011520817.1 | 7, 8 | 1122 | |||
XM_011522516.2 | XP_011520818.1 | 6, 7 | 882 | |||
XM_017023268.1 | XP_016878757.1 | 7, 8 | 1119 | |||
GenBank mRNA | AK027646.1 | - | 6, 7 | 959 | ||
AK297094.1 | - | 6, 7 | 513 | |||
BC002856.1 | - | 7, 8 | 1095 | 3635977 |