Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YARS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040436.2 | NP_001035526.1 | 2 | 968 | ||
GenBank mRNA | AF132939.1 | - | 2 | 891 | ||
AK024057.1 | - | 2 | 916 | |||
AK026619.1 | - | 2 | 915 | |||
AK130935.1 | - | 3 | 852 | |||
BC015625.2 | - | 2 | 891 | 4843916 |