Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NMD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320227.1 | NP_001307156.1 | 16 | 1591 | ||
NM_015938.4 | NP_057022.2 | 16 | 1591 | |||
XM_005247511.1 | XP_005247568.1 | 16 | 1541 | |||
XM_005247512.1 | XP_005247569.1 | 16 | 1872 | |||
GenBank mRNA | AF132941.1 | - | 16 | 1562 | ||
AK021775.1 | - | 13 | 1202 | |||
AK024706.1 | - | 3 | 192 | |||
AK094749.1 | - | 15 | 1894 | |||
AK225664.1 | - | 16 | 1573 | |||
AK296710.1 | - | 15 | 1199 | |||
BC013317.1 | - | 16 | 1574 | 4291797 |