Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIDT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040455.1 | NP_001035545.1 | 13 | 1212 | ||
GenBank mRNA | AF151799.1 | - | 14 | 1317 | ||
AK075137.1 | - | 4 | 190 | |||
AK292501.1 | - | 13 | 1692 | |||
AK292974.1 | - | 13 | 1203 | |||
AK301501.1 | - | 5 | 482 | |||
AK303023.1 | - | 13 | 1168 | |||
AY358442.1 | - | 13 | 1221 | |||
BX648437.1 | - | 5 | 310 | |||
KC109742.1 | - | 13 | 1168 |