Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADIPOR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290553.1 | NP_001277482.1 | 4, 5 | 590 | ||
NM_001290557.1 | NP_001277486.1 | 5, 6 | 793 | |||
NM_001290629.1 | NP_001277558.1 | 5, 6 | 679 | |||
NM_015999.5 | NP_057083.2 | 4, 5 | 726 | |||
XM_017001407.1 | XP_016856896.1 | 5, 6 | 677 | |||
GenBank mRNA | AF125179.1 | - | 4, 5 | 594 | ||
AF151803.1 | - | 4, 5 | 602 | |||
AK001484.1 | - | 4, 5 | 642 | |||
AK058114.1 | - | 4, 5 | 642 | |||
AK222503.1 | - | 4, 5 | 600 | |||
AY424279.1 | - | 4, 5 | 562 | |||
BC001594.2 | - | 4, 5 | 619 | 2988737 | ||
BC010743.1 | - | 4, 5 | 613 | 3878067 | ||
BI597715.1 | - | 5, 6 | 682 | |||
BX461217.2 | - | 5, 6 | 679 |