Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADIPOR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290553.1 | NP_001277482.1 | 5 | 739 | ||
NM_001290557.1 | NP_001277486.1 | 6 | 942 | |||
NM_001290629.1 | NP_001277558.1 | 6 | 828 | |||
NM_015999.5 | NP_057083.2 | 5 | 875 | |||
XM_017001407.1 | XP_016856896.1 | 6 | 852 | |||
GenBank mRNA | AF125179.1 | - | 5 | 743 | ||
AF151803.1 | - | 5 | 751 | |||
AK001484.1 | - | 5 | 791 | |||
AK058114.1 | - | 5 | 791 | |||
AK222503.1 | - | 5 | 749 | |||
AY424279.1 | - | 5 | 711 | |||
BC001594.2 | - | 5 | 768 | 2988737 | ||
BC010743.1 | - | 5 | 762 | 3878067 | ||
BX461217.2 | - | 6 | 828 |