Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UTP18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016001.2 | NP_057085.2 | 8 | 1127 | ||
XM_006721930.3 | XP_006721993.2 | 8 | 1106 | |||
XM_011524870.2 | XP_011523172.1 | 8 | 1106 | |||
GenBank mRNA | AF151806.1 | - | 8 | 1113 | ||
AK026355.1 | - | 9 | 876 | |||
AK314399.1 | - | 8 | 1106 | |||
AY007138.1 | - | 6 | 597 | |||
BC025276.1 | - | 8 | 1103 | 4861495 | ||
BM548967.1 | - | 7 | 763 |