Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFT52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303458.2 | NP_001290387.1 | 3, 4 | 635 | ||
NM_001303459.2 | NP_001290388.1 | 3, 4 | 635 | |||
NM_001323578.1 | NP_001310507.1 | 3, 4 | 660 | |||
NM_001323579.1 | NP_001310508.1 | 3, 4 | 660 | |||
NM_001323580.1 | NP_001310509.1 | 3, 4 | 635 | |||
NM_001323581.1 | NP_001310510.1 | 3, 4 | 635 | |||
NM_016004.4 | NP_057088.2 | 3, 4 | 660 | |||
XM_017027863.1 | XP_016883352.1 | 3, 4 | 291 | |||
GenBank mRNA | AF151811.1 | - | 3, 4 | 257 | ||
AK001436.1 | - | 3, 4 | 260 | |||
BC039831.1 | - | 3, 4 | 341 | 6095634 | ||
BP308189.1 | - | 3, 4 | 318 |