Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH3GLB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206651.1 | NP_001193580.1 | 2 | 467 | ||
NM_001206652.1 | NP_001193581.1 | 2 | 467 | |||
NM_016009.4 | NP_057093.1 | 2 | 467 | |||
XM_006710672.2 | XP_006710735.1 | 2 | 467 | |||
XM_011541538.1 | XP_011539840.1 | 2 | 467 | |||
XM_017001409.1 | XP_016856898.1 | 2 | 467 | |||
XM_017001410.1 | XP_016856899.1 | 2 | 467 | |||
GenBank mRNA | AB007960.2 | - | 2 | 432 | ||
AF151819.1 | - | 2 | 162 | |||
AF257318.1 | - | 2 | 162 | |||
AF263293.1 | - | 2 | 142 | |||
AF350371.1 | - | 2 | 218 | |||
AK001954.1 | - | 2 | 252 | |||
AK225864.1 | - | 2 | 287 | |||
BC007455.2 | - | 2 | 182 | 2988769 | ||
DB038227.1 | - | 2 | 467 | |||
DB451777.1 | - | 2 | 463 |