Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COQ4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016035.4 | NP_057119.2 | 6 | 948 | ||
XM_017014792.1 | XP_016870281.1 | 4 | 743 | |||
GenBank mRNA | AF151850.1 | - | 6 | 919 | ||
AK025376.1 | - | 6 | 862 | |||
AK095083.1 | - | 2 | 2413 | |||
AK128853.1 | - | 1 | 4612 | |||
AK313650.1 | - | 6 | 931 | |||
BC011895.2 | - | 6 | 618 | 3944239 | ||
EU216419.1 | - | 6 | 930 | |||
EU216420.1 | - | 6 | 848 | |||
EU216421.1 | - | 6 | 682 | |||
EU216422.1 | - | 6 | 620 | |||
EU216423.1 | - | 5 | 544 |