Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCTD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319294.1 | NP_001306223.1 | 16 | 1892 | ||
NM_001319295.1 | NP_001306224.1 | 16 | 1964 | |||
NM_016121.4 | NP_057205.2 | 16 | 1892 | |||
XM_005273158.1 | XP_005273215.1 | 16 | 2158 | |||
GenBank mRNA | AK055925.1 | - | 10 | 1085 | ||
AK296275.1 | - | 14 | 1425 | |||
AK316272.1 | - | 12 | 1293 | |||
AL833887.1 | - | 2 | 86 | |||
AL834389.1 | - | 10 | 1116 | |||
BC013868.2 | - | 16 | 1879 | |||
BC037896.1 | - | 1 | 669 | |||
BC094879.1 | - | 16 | 1841 | 3078971 | ||
BC117188.1 | - | 16 | 1774 | 40125739 | ||
BC126173.1 | - | 16 | 1774 | 8991889 |