Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INSIG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321329.1 | NP_001308258.1 | 2 | 334 | ||
NM_016133.3 | NP_057217.2 | 2 | 341 | |||
GenBank mRNA | AF125392.1 | - | 1 | 255 | ||
AF527632.1 | - | 2 | 290 | |||
AK291433.1 | - | 2 | 289 | |||
BC022475.1 | - | 2 | 285 | 4794938 | ||
HY053967.1 | - | 2 | 343 |