Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DYNC1LI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016141.3 | NP_057225.2 | 2, 3 | 315 | ||
GenBank mRNA | AF078849.1 | - | 2, 3 | 291 | ||
AK074348.1 | - | 2, 3 | 299 | |||
AK222573.1 | - | 2, 3 | 250 | |||
AK222653.1 | - | 2, 3 | 251 | |||
BC131620.1 | - | 2, 3 | 261 | 40117391 | ||
BX649189.1 | - | 2, 3 | 134 | |||
DA284201.1 | - | 2, 3 | 315 | |||
HY036998.1 | - | 2, 3 | 368 |