Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AADAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286682.1 | NP_001273611.1 | 13 | 1293 | ||
NM_001286683.1 | NP_001273612.1 | 13 | 1366 | |||
NM_016228.3 | NP_057312.1 | 12 | 1499 | |||
NM_182662.1 | NP_872603.1 | 13 | 1281 | |||
XM_006714231.2 | XP_006714294.1 | 14 | 1423 | |||
XM_011532020.2 | XP_011530322.1 | 10 | 1056 | |||
GenBank mRNA | AF481738.1 | - | 12 | 1500 | ||
AI190763.1 | - | 3 | 338 | |||
AK055952.1 | - | 13 | 1281 | |||
AK314729.1 | - | 13 | 1368 | |||
BC031068.1 | - | 13 | 1286 | 5286180 |