Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCXR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195218.1 | NP_001182147.1 | 5, 6 | 453 | ||
NM_016286.3 | NP_057370.1 | 5, 6 | 459 | |||
GenBank mRNA | AA446582.1 | - | 3, 4 | 372 | ||
AB013846.1 | - | 5, 6 | 444 | |||
AF113123.1 | - | 5, 6 | 447 | |||
AF139841.1 | - | 5, 6 | 451 | |||
AF515623.1 | - | 5, 6 | 444 | |||
AF515624.1 | - | 5, 6 | 444 | |||
AF515625.1 | - | 5, 6 | 444 | |||
AK226040.1 | - | 5, 6 | 465 | |||
BC001470.2 | - | 5, 6 | 445 | 3138707 | ||
BC003018.1 | - | 5, 6 | 442 | |||
BG434209.1 | - | 5, 6 | 458 | |||
BT006881.1 | - | 5, 6 | 444 | |||
CB959852.1 | - | 5, 6 | 486 |