Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDKL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113575.1 | NP_001107047.1 | 3 | 415 | ||
NM_001300853.1 | NP_001287782.1 | 2 | 181 | |||
NM_016508.3 | NP_057592.2 | 3 | 415 | |||
XM_011543429.2 | XP_011541731.1 | 3 | 730 | |||
XM_017009524.1 | XP_016865013.1 | 2 | 1162 | |||
XM_017009525.1 | XP_016865014.1 | 3 | 730 | |||
XM_017009526.1 | XP_016865015.1 | 3 | 728 | |||
XM_017009527.1 | XP_016865016.1 | 2 | 1162 | |||
XM_017009528.1 | XP_016865017.1 | 2 | 1162 | |||
XM_017009529.1 | XP_016865018.1 | 2 | 1162 | |||
XM_017009530.1 | XP_016865019.1 | 2 | 1162 | |||
XM_017009531.1 | XP_016865020.1 | 2 | 1162 | |||
XM_017009532.1 | XP_016865021.1 | 2 | 1161 | |||
XM_017009533.1 | XP_016865022.1 | 2 | 1161 | |||
XM_017009534.1 | XP_016865023.1 | 2 | 1161 | |||
XM_017009535.1 | XP_016865024.1 | 2 | 1161 | |||
XM_017009536.1 | XP_016865025.1 | 2 | 1163 | |||
XM_017009538.1 | XP_016865027.1 | 2 | 206 | |||
XM_017009539.1 | XP_016865028.1 | 2 | 206 | |||
XM_017009541.1 | XP_016865030.1 | 2 | 542 | |||
XM_017009544.1 | XP_016865033.1 | 2 | 206 | |||
XM_017009548.1 | XP_016865037.1 | 2 | 1166 | |||
GenBank mRNA | AF130372.1 | - | 2 | 296 | ||
AK301802.1 | - | 2 | 175 | |||
BC041799.1 | - | 3 | 417 | 5298279 |