Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM53C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135647.1 | NP_001129119.1 | 4 | 1000 | ||
NM_016605.2 | NP_057689.1 | 4 | 975 | |||
GenBank mRNA | AF251040.1 | - | 4 | 711 | ||
AK315567.1 | - | 4 | 724 | |||
BC000259.1 | - | 4 | 717 | 3357346 | ||
BC052993.1 | - | 4 | 940 | 5164799 |