Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NME8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016616.4 | NP_057700.3 | 15 | 1669 | ||
GenBank mRNA | AF202051.2 | - | 14 | 1398 | ||
AF305596.1 | - | 14 | 1367 | |||
AK313599.1 | - | 14 | 1409 | |||
BC036816.1 | - | 15 | 1669 | 5745079 | ||
GQ472221.1 | - | 15 | 1669 |