Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RSRC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271834.1 | NP_001258763.1 | 9 | 934 | ||
NM_001271838.1 | NP_001258767.1 | 10 | 1134 | |||
NM_016625.3 | NP_057709.2 | 10 | 1122 | |||
GenBank mRNA | AK290334.1 | - | 10 | 1102 | ||
BC006982.1 | - | 10 | 1071 | 3997840 | ||
BC010357.1 | - | 9 | 883 | 4249317 |