Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NGRN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033088.1 | NP_001028260.2 | 3 | 585 | ||
NR_028052.1 | 2 | 996 | ||||
GenBank mRNA | AB029315.1 | - | 2 | 994 | ||
AF225423.1 | - | 2 | 885 | |||
AF242770.1 | - | 2 | 950 | |||
AK312641.1 | - | 3 | 799 | |||
AL360142.1 | - | 2 | 968 | |||
AY049780.1 | - | 2 | 319 | |||
BC001682.2 | - | 3 | 567 | 2964457 | ||
BC007222.1 | - | 2 | 971 | 2819984 | ||
BC009389.2 | - | 2 | 946 | 4130266 | ||
BC017192.2 | - | 2 | 951 | 3139180 | ||
BC019023.2 | - | 2 | 952 | |||
BC096824.1 | - | 3 | 502 | |||
CD242640.1 | - | 3 | 572 |