Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AIG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286587.1 | NP_001273516.1 | 6 | 639 | ||
NM_016108.3 | NP_057192.2 | 5 | 696 | |||
XM_011535881.2 | XP_011534183.1 | 4 | 562 | |||
XM_011535882.2 | XP_011534184.1 | 4 | 561 | |||
XM_011535883.2 | XP_011534185.1 | 5 | 677 | |||
XM_011535884.2 | XP_011534186.1 | 5 | 679 | |||
XM_011535885.2 | XP_011534187.1 | 4 | 561 | |||
XM_017010927.1 | XP_016866416.1 | 5 | 695 | |||
XM_017010928.1 | XP_016866417.1 | 4 | 561 | |||
GenBank mRNA | AF151861.1 | - | 6 | 645 | ||
AF153605.1 | - | 5 | 639 | |||
AJ420480.1 | - | 3 | 312 | |||
AK001347.1 | - | 5 | 637 | |||
AK222475.1 | - | 5 | 658 | |||
BC025278.1 | - | 5 | 627 | 4859216 | ||
BX538067.1 | - | 4 | 399 |