Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM14C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165258.1 | NP_001158730.1 | 6 | 696 | ||
NM_016462.3 | NP_057546.1 | 6 | 682 | |||
GenBank mRNA | AF151028.1 | - | 6 | 492 | ||
AF164962.1 | - | 5 | 427 | |||
AK223462.1 | - | 6 | 380 | |||
AL702149.1 | - | 6 | 516 | |||
BC002496.2 | - | 6 | 417 | 3050609 | ||
BC010086.2 | - | 6 | 446 | 2907072 | ||
CX784985.1 | - | 6 | 682 |